Assay Details
| Gene Symbol: Slc44a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199186.1 | NP_001186115.1 | 7 - 8 | 596 | 64 | |
| NM_152808.3 | NP_690021.1 | 7 - 8 | 598 | 64 | ||
| GenBank mRNA | AK029457.1 | - | 7 - 8 | 558 | 64 | |
| AK041533.1 | - | 7 - 8 | 555 | 64 | ||
| AK043924.1 | - | 7 - 8 | 568 | 64 | ||
| AK048648.1 | - | 7 - 8 | 583 | 64 | ||
| AK088553.1 | - | 7 - 8 | 545 | 64 | ||
| AK153992.1 | - | 7 - 8 | 577 | 64 | ||
| AK166695.1 | - | 7 - 8 | 599 | 64 | ||
| BC023763.1 | - | 7 - 8 | 581 | 64 | ||
| BC031535.1 | - | 7 - 8 | 574 | 3991609 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc44a2
Genomic Map