Assay Details
| Gene Symbol: Cdc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016742.4 | NP_058022.1 | 1 - 2 | 155 | 87 | |
| GenBank mRNA | AK010494.1 | - | 1 - 2 | 180 | 87 | |
| AK013255.1 | - | 1 - 2 | 156 | 87 | ||
| AK145671.1 | - | 1 - 2 | 187 | 87 | ||
| AK168651.1 | - | 1 - 2 | 173 | 87 | ||
| BC060079.1 | - | 1 - 2 | 178 | 5714770 | 87 | |
| U43076.1 | - | 1 - 2 | 193 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdc37
Genomic Map