Assay Details
| Gene Symbol: Cdc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016742.4 | NP_058022.1 | 3 - 4 | 541 | 101 | |
| GenBank mRNA | AK010494.1 | - | 3 - 4 | 566 | 101 | |
| AK013255.1 | - | 3 - 4 | 542 | 101 | ||
| AK145671.1 | - | 3 - 4 | 573 | 101 | ||
| AK168651.1 | - | 3 - 4 | 559 | 101 | ||
| BC060079.1 | - | 3 - 4 | 564 | 5714770 | 101 | |
| U43076.1 | - | 3 - 4 | 579 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdc37
Genomic Map