Assay Details
| Gene Symbol: Taf1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028401.1 | 8 - 9 | 1056 | 79 | ||
| GenBank mRNA | AK012656.1 | - | 6 - 7 | 905 | 79 | |
| AK014920.1 | - | 8 - 9 | 1008 | 79 | ||
| AK020036.1 | - | 7 - 8 | 968 | 79 | ||
| AK050165.1 | - | 8 - 9 | 1012 | 79 | ||
| AK166329.1 | - | 8 - 9 | 1011 | 79 | ||
| BC056964.1 | - | 5 - 6 | 3227 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Taf1d
Genomic Map