Assay Details
| Gene Symbol: Kank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145611.4 | NP_663586.3 | 1 - 2 | 63 | 56 | |
| GenBank mRNA | AK045492.1 | - | 1 - 2 | 35 | 56 | |
| AK049288.1 | - | 1 - 2 | 64 | 56 | ||
| BC006647.1 | - | 1 - 2 | 50 | 56 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK045492.1, AK049288.1, NM_145611.4 transcripts.
Set Membership: |
|

Kank2
Genomic Map