Assay Details
| Gene Symbol: Kank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145611.4 | NP_663586.3 | 4 - 5 | 1544 | 66 | |
| GenBank mRNA | AK031324.1 | - | 4 - 5 | 1537 | 66 | |
| AK045492.1 | - | 4 - 5 | 1516 | 66 | ||
| AK049288.1 | - | 4 - 5 | 1545 | 66 | ||
| BC006647.1 | - | 4 - 5 | 1532 | 66 | ||
| BC010245.1 | - | 4 - 5 | 1501 | 3710765 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kank2
Genomic Map