Assay Details
| Gene Symbol: Prkcsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293650.1 | NP_001280579.1 | 16 - 17 | 1598 | 72 | |
| NM_001293651.1 | NP_001280580.1 | 16 - 17 | 1581 | 72 | ||
| NM_008925.2 | NP_032951.1 | 16 - 17 | 1577 | 72 | ||
| GenBank mRNA | AK137661.1 | - | 16 - 17 | 1544 | 72 | |
| AK153931.1 | - | 16 - 17 | 1552 | 72 | ||
| AK159705.1 | - | 16 - 17 | 1548 | 72 | ||
| BC009816.1 | - | 16 - 17 | 1531 | 4008182 | 72 | |
| U92794.1 | - | 16 - 17 | 1530 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkcsh
Genomic Map