Assay Details
| Gene Symbol: Rgl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023622.4 | NP_076111.2 | 16 - 17 | 1797 | 58 | |
| GenBank mRNA | AF237669.1 | - | 16 - 17 | 1756 | 58 | |
| AF239661.1 | - | 16 - 17 | 1798 | 58 | ||
| AK004876.1 | - | 13 - 14 | 1825 | 58 | ||
| AK053550.1 | - | 16 - 17 | 1784 | 58 | ||
| AK131140.1 | - | 1303 | 58 | |||
| AK134654.1 | - | 1779 | 58 | |||
| AK165023.1 | - | 13 - 14 | 2186 | 58 | ||
| AK165746.1 | - | 14 - 15 | 2088 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rgl3
Genomic Map