Assay Details
| Gene Symbol: Ldlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252658.1 | NP_001239587.1 | 2 - 3 | 396 | 63 | |
| NM_001252659.1 | NP_001239588.1 | 2 - 3 | 393 | 63 | ||
| NM_010700.3 | NP_034830.2 | 2 - 3 | 396 | 63 | ||
| GenBank mRNA | AF425607.1 | - | 2 - 3 | 244 | 63 | |
| AK076243.1 | - | 2 - 3 | 374 | 63 | ||
| AK149950.1 | - | 2 - 3 | 334 | 63 | ||
| AK151968.1 | - | 2 - 3 | 363 | 63 | ||
| AK152103.1 | - | 2 - 3 | 332 | 63 | ||
| AK160006.1 | - | 2 - 3 | 334 | 63 | ||
| AK161104.1 | - | 2 - 3 | 334 | 63 | ||
| AK166466.1 | - | 2 - 3 | 333 | 63 | ||
| AK170264.1 | - | 2 - 3 | 341 | 63 | ||
| BC019207.1 | - | 2 - 3 | 314 | 5052599 | 63 | |
| BC053041.1 | - | 2 - 3 | 341 | 5704871 | 63 | |
| X64414.1 | - | 2 - 3 | 315 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ldlr
Genomic Map