Assay Details
| Gene Symbol: Ldlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252658.1 | NP_001239587.1 | 3 - 4 | 510 | 78 | |
| NM_001252659.1 | NP_001239588.1 | 3 - 4 | 507 | 78 | ||
| NM_010700.3 | NP_034830.2 | 3 - 4 | 510 | 78 | ||
| GenBank mRNA | AF425607.1 | - | 3 - 4 | 358 | 78 | |
| AK018697.1 | - | 3 - 4 | 446 | 78 | ||
| AK076243.1 | - | 3 - 4 | 488 | 78 | ||
| AK149950.1 | - | 3 - 4 | 448 | 78 | ||
| AK151968.1 | - | 3 - 4 | 477 | 78 | ||
| AK152103.1 | - | 3 - 4 | 446 | 78 | ||
| AK160006.1 | - | 3 - 4 | 448 | 78 | ||
| AK161104.1 | - | 3 - 4 | 448 | 78 | ||
| AK170264.1 | - | 3 - 4 | 455 | 78 | ||
| BC019207.1 | - | 3 - 4 | 428 | 5052599 | 78 | |
| BC053041.1 | - | 3 - 4 | 455 | 5704871 | 78 | |
| X64414.1 | - | 3 - 4 | 429 | 78 | ||
| Z19521.1 | - | 3 - 4 | 308 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ldlr
Genomic Map