Assay Details
| Gene Symbol: Ldlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252658.1 | NP_001239587.1 | 5 - 6 | 1024 | 93 | |
| NM_001252659.1 | NP_001239588.1 | 5 - 6 | 1021 | 93 | ||
| NM_010700.3 | NP_034830.2 | 5 - 6 | 1024 | 93 | ||
| GenBank mRNA | AK018697.1 | - | 5 - 6 | 956 | 93 | |
| AK076243.1 | - | 5 - 6 | 1002 | 93 | ||
| AK149950.1 | - | 5 - 6 | 962 | 93 | ||
| AK151968.1 | - | 5 - 6 | 991 | 93 | ||
| AK152103.1 | - | 5 - 6 | 960 | 93 | ||
| AK160006.1 | - | 5 - 6 | 962 | 93 | ||
| AK161104.1 | - | 5 - 6 | 962 | 93 | ||
| AK166466.1 | - | 5 - 6 | 961 | 93 | ||
| BC053041.1 | - | 5 - 6 | 969 | 5704871 | 93 | |
| X64414.1 | - | 5 - 6 | 943 | 93 | ||
| Z19521.1 | - | 5 - 6 | 822 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ldlr
Genomic Map