Assay Details
| Gene Symbol: Hepacam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175189.4 | NP_780398.2 | 1 - 2 | 157 | 69 | |
| GenBank mRNA | AK013637.1 | - | 1 - 2 | 69 | ||
| AK081154.1 | - | 1 - 2 | 73 | 69 | ||
| AK158265.1 | - | 1 - 2 | 395 | 69 | ||
| BC082537.1 | - | 1 - 2 | 71 | 69 | ||
| BC139057.1 | - | 1 - 2 | 147 | 8862079 | 69 | |
| BC139058.1 | - | 1 - 2 | 147 | 8862080 | 69 | |
| BY753058.1 | - | 1 - 2 | 392 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hepacam
Genomic Map