Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900019841.mm.1 |
NM_176831.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900019841.mm.1 |
NM_176831.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900019841.mm.1 |
TC0900002368.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900019844.mm.1 |
NM_176831.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900019844.mm.1 |
NM_176831.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900019844.mm.1 |
TC0900002368.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900019839.mm.1 |
NM_176831.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900019839.mm.1 |
NM_176831.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900019839.mm.1 |
TC0900002368.mm.1 |
MTA TC |