Assay Details
| Gene Symbol: Cadm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025600.1 | NP_001020771.1 | 2 - 3 | 301 | 103 | |
| NM_018770.3 | NP_061240.3 | 2 - 3 | 301 | 103 | ||
| NM_207675.2 | NP_997558.2 | 2 - 3 | 301 | 103 | ||
| NM_207676.2 | NP_997559.1 | 2 - 3 | 301 | 103 | ||
| GenBank mRNA | AB052293.1 | - | 2 - 3 | 301 | 103 | |
| AB064265.2 | - | 2 - 3 | 280 | 103 | ||
| AB092414.1 | - | 2 - 3 | 280 | 103 | ||
| AB183399.1 | - | 2 - 3 | 280 | 103 | ||
| AB183400.1 | - | 2 - 3 | 280 | 103 | ||
| AB183401.1 | - | 2 - 3 | 280 | 103 | ||
| AB183402.1 | - | 2 - 3 | 280 | 103 | ||
| AB377192.1 | - | 2 - 3 | 301 | 103 | ||
| AF061260.1 | - | 1 - 2 | 138 | 103 | ||
| AF434663.1 | - | 2 - 3 | 396 | 103 | ||
| AF539424.1 | - | 2 - 3 | 293 | 103 | ||
| AK013775.1 | - | 2 - 3 | 302 | 103 | ||
| AK013911.1 | - | 2 - 3 | 395 | 103 | ||
| AK053839.1 | - | 2 - 3 | 302 | 103 | ||
| AY351388.1 | - | 2 - 3 | 280 | 103 | ||
| BC057125.1 | - | 2 - 3 | 289 | 6414884 | 103 | |
| BC090970.1 | - | 2 - 3 | 480 | 6703838 | 103 | |
| BC095986.1 | - | 2 - 3 | 293 | 5372277 | 103 | |
| DQ279856.1 | - | 2 - 3 | 280 | 103 | ||
| JQ782219.1 | - | 2 - 3 | 280 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF061260.1 transcripts.
Set Membership: |
|

Cadm1
Genomic Map