Assay Details
| Gene Symbol: Cadm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207675.2 | NP_997558.2 | 9 - 10 | 1140 | 61 | |
| NM_207676.2 | NP_997559.1 | 8 - 9 | 1056 | 61 | ||
| GenBank mRNA | AB021964.1 | - | 6 - 7 | 636 | 61 | |
| AB021965.1 | - | 6 - 7 | 618 | 61 | ||
| AB021966.1 | - | 6 - 7 | 669 | 61 | ||
| AB021967.1 | - | 5 - 6 | 585 | 61 | ||
| AB064265.2 | - | 9 - 10 | 1119 | 61 | ||
| AB183400.1 | - | 8 - 9 | 1035 | 61 | ||
| AK013911.1 | - | 9 - 10 | 1234 | 61 | ||
| BC057125.1 | - | 8 - 9 | 1044 | 6414884 | 61 | |
| BC090970.1 | - | 8 - 9 | 1235 | 6703838 | 61 | |
| BC094661.1 | - | 5 - 6 | 606 | 61 | ||
| DQ279856.1 | - | 8 - 9 | 1035 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cadm1
Genomic Map