Assay Details
| Gene Symbol: Car12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052639.1 | - | 3 - 4 | 405 | 69 | |
| AK157977.1 | - | 3 - 4 | 405 | 69 | ||
| AK166132.1 | - | 3 - 4 | 403 | 69 | ||
| AK170859.1 | - | 3 - 4 | 364 | 69 | ||
| BC031385.1 | - | 3 - 4 | 389 | 5355557 | 69 | |
| BC033432.1 | - | 3 - 4 | 314 | 69 | ||
| BC035941.1 | - | 3 - 4 | 389 | 3987392 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car12
Genomic Map