Assay Details
| Gene Symbol: Car12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052639.1 | - | 7 - 8 | 870 | 64 | |
| AK157977.1 | - | 7 - 8 | 870 | 64 | ||
| AK166132.1 | - | 7 - 8 | 868 | 64 | ||
| AK170859.1 | - | 7 - 8 | 829 | 64 | ||
| BC031385.1 | - | 7 - 8 | 854 | 5355557 | 64 | |
| BC033432.1 | - | 7 - 8 | 779 | 64 | ||
| BC035941.1 | - | 7 - 8 | 854 | 3987392 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car12
Genomic Map