Assay Details
| Gene Symbol: Naalad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028279.3 | NP_082555.2 | 8 - 9 | 1040 | 125 | |
| GenBank mRNA | AK133611.1 | - | 8 - 9 | 1150 | 125 | |
| AK135653.1 | - | 10 - 11 | 1183 | 125 | ||
| AK161457.1 | - | 11 - 12 | 1346 | 125 | ||
| AY243507.1 | - | 8 - 9 | 1040 | 125 | ||
| BC108995.1 | - | 8 - 9 | 1024 | 40047859 | 125 | |
| BC108996.1 | - | 8 - 9 | 1024 | 40047860 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Naalad2
Genomic Map