Assay Details
| Gene Symbol: Naalad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028279.3 | NP_082555.2 | 12 - 13 | 1392 | 97 | |
| GenBank mRNA | AK133611.1 | - | 12 - 13 | 1502 | 97 | |
| AK135653.1 | - | 14 - 15 | 1535 | 97 | ||
| AK161457.1 | - | 15 - 16 | 1698 | 97 | ||
| AY243507.1 | - | 12 - 13 | 1392 | 97 | ||
| BC108995.1 | - | 12 - 13 | 1376 | 40047859 | 97 | |
| BC108996.1 | - | 12 - 13 | 1376 | 40047860 | 97 | |
More Information
Set Membership: |
|

Naalad2
Genomic Map