Assay Details
| Gene Symbol: Naalad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028279.3 | NP_082555.2 | 2 - 3 | 249 | 152 | |
| GenBank mRNA | AK133611.1 | - | 2 - 3 | 359 | 152 | |
| AK135653.1 | - | 4 - 5 | 391 | 152 | ||
| AK161457.1 | - | 5 - 6 | 555 | 152 | ||
| AY243507.1 | - | 2 - 3 | 249 | 152 | ||
| BC108995.1 | - | 2 - 3 | 233 | 40047859 | 152 | |
| BC108996.1 | - | 2 - 3 | 233 | 40047860 | 152 | |
More Information
Set Membership: |
|

Naalad2
Genomic Map