Assay Details
| Gene Symbol: Parp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177460.4 | NP_803411.3 | 3 - 4 | 590 | 63 | |
| GenBank mRNA | AK033791.1 | - | 540 | 63 | ||
| AK133475.1 | - | 3 - 4 | 591 | 63 | ||
| AK166002.1 | - | 2 - 3 | 354 | 63 | ||
| AK169866.1 | - | 3 - 4 | 505 | 63 | ||
| BC014731.1 | - | 2 - 3 | 620 | 63 | ||
| BC055447.1 | - | 3 - 4 | 559 | 3594406 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parp16
Genomic Map