Assay Details
| Gene Symbol: Parp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177460.4 | NP_803411.3 | 5 - 6 | 980 | 75 | |
| GenBank mRNA | AK033791.1 | - | 930 | 75 | ||
| AK133475.1 | - | 5 - 6 | 981 | 75 | ||
| BC002197.1 | - | 1 - 2 | 123 | 75 | ||
| BC014731.1 | - | 4 - 5 | 1010 | 75 | ||
| BC055447.1 | - | 5 - 6 | 949 | 3594406 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parp16
Genomic Map