Assay Details
| Gene Symbol: Atg4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153583.10 | NP_705811.8 | 2 - 3 | 520 | 77 | |
| GenBank mRNA | AK036282.1 | - | 2 - 3 | 507 | 77 | |
| AK036571.1 | - | 2 - 3 | 509 | 77 | ||
| AK081002.1 | - | 2 - 3 | 507 | 77 | ||
| AK081605.1 | - | 2 - 3 | 498 | 77 | ||
| AK087258.1 | - | 2 - 3 | 510 | 77 | ||
| AK154130.1 | - | 2 - 3 | 77 | |||
| BC030861.1 | - | 2 - 3 | 4208700 | 77 | ||
| BC069851.1 | - | 2 - 3 | 472 | 6490007 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atg4d
Genomic Map