Assay Details
| Gene Symbol: Zfp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011753.3 | NP_035883.2 | 2 - 3 | 330 | 94 | |
| GenBank mRNA | AK017201.1 | - | 2 - 3 | 160 | 94 | |
| AK141740.1 | - | 2 - 3 | 167 | 94 | ||
| CA871137.1 | - | 2 - 3 | 330 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK141740.1, NM_011753.3 transcripts.
Set Membership: |
|

Zfp26
Genomic Map