Assay Details
| Gene Symbol: Unc13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081153.1 | NP_001074622.1 | 27 - 28 | 5867 | 73 | |
| GenBank mRNA | AK134584.1 | - | 1 - 2 | 64 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Unc13c
Genomic Map