Assay Details
| Gene Symbol: Unc13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081153.1 | NP_001074622.1 | 23 - 24 | 5448 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc13c| Gene Symbol: Unc13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081153.1 | NP_001074622.1 | 23 - 24 | 5448 | 100 | |
Set Membership: |
|