Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000006444.mm.1 |
NM_001122604.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006444.mm.1 |
NM_001122604.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006444.mm.1 |
NM_001122605.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006444.mm.1 |
NM_001122605.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006444.mm.1 |
NM_133188.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006444.mm.1 |
NM_133188.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006444.mm.1 |
TC1000000848.mm.1 |
MTA TC |