Assay Details
| Gene Symbol: Psmd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026545.3 | NP_080821.3 | 5 - 6 | 840 | 75 | |
| GenBank mRNA | AK002921.1 | - | 5 - 6 | 679 | 75 | |
| AK003436.1 | - | 5 - 6 | 681 | 75 | ||
| AK027973.1 | - | 5 - 6 | 644 | 75 | ||
| AK150493.1 | - | 5 - 6 | 683 | 75 | ||
| AK150982.1 | - | 5 - 6 | 647 | 75 | ||
| AK159216.1 | - | 5 - 6 | 649 | 75 | ||
| AK159795.1 | - | 5 - 6 | 665 | 75 | ||
| AK159926.1 | - | 5 - 6 | 675 | 75 | ||
| AK166718.1 | - | 5 - 6 | 674 | 75 | ||
| AK167948.1 | - | 5 - 6 | 649 | 75 | ||
| AK168886.1 | - | 5 - 6 | 648 | 75 | ||
| BC004075.1 | - | 5 - 6 | 658 | 3591796 | 75 | |
| BC005717.1 | - | 5 - 6 | 568 | 2609781 | 75 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25025802 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25025804 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25025803 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P149325 | AK020132.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P149325 | AK020132.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 788008 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 292724 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527968 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 185364 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 36817 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140256 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P149325 | AK020132.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P149325 | AK020132.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P149325 | AK020132.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P149325 | AK020132.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25025819 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140256 | - | - |
| Whole Mouse Genome G4122A | A_51_P149325 | AK020132.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P149325 | AK020132.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126912 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127911 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140256 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06094596 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06094589 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06094600 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06094593 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06094595 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25025804 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25025802 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25025803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25025757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06117757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06264526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06094595 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05982233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06149361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06227039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06642031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06149359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25025775 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10561688 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561682 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561684 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561681 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561687 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561683 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561689 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561680 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561686 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561685 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423296_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10561679 | - | - |
| Mouse Genome 430 2.0 Array | 1423296_at | - | - |
| Mouse Genome 430 2.0 Array | 1423296_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57296_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57296_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700023958.mm.1 | TC0700002716.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700023958.mm.1 | NM_026545.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700023958.mm.1 | NM_026545.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700023956.mm.1 | TC0700002716.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700023956.mm.1 | NM_026545.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700023956.mm.1 | NM_026545.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700023954.mm.1 | TC0700002716.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700023954.mm.1 | NM_026545.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700023954.mm.1 | NM_026545.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700023960.mm.1 | TC0700002716.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700023960.mm.1 | NM_026545.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700023960.mm.1 | NM_026545.3 | Refseq |
| Mouse Expression Set 430 | 1423296_at | - | - |
| Mouse Expression Set 430 | 1423296_PM_at | - | - |
| Murine Genome U74v2 Set | 98522_at | - | - |
| Murine Genome U74v2 Set | 98522_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423296_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423296_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423296_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423296_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102360647 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 630142 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000630142 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2678019 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2546047 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2678019 | - | - |
| mouse-6 v1.1 (Illumina) | scl0057296.1_256-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 630142 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0057296.1_256-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 630142 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2678019 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2678019 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0057296.1_256-S | - | - |

Psmd8
Genomic Map