Assay Details
| Gene Symbol: Fam118b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286604.1 | NP_001273533.1 | 4 - 5 | 310 | 121 | |
| NM_175411.5 | NP_780620.1 | 2 - 3 | 180 | 121 | ||
| NM_194257.2 | NP_919233.1 | 2 - 3 | 180 | 121 | ||
| GenBank mRNA | AK079397.1 | - | 4 - 5 | 283 | 121 | |
| AK081206.1 | - | 2 - 3 | 156 | 121 | ||
| AK168168.1 | - | 2 - 3 | 156 | 121 | ||
| AK169810.1 | - | 2 - 3 | 155 | 121 | ||
| BC051180.1 | - | 2 - 3 | 142 | 6315210 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam118b
Genomic Map