Assay Details
| Gene Symbol: Gns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029364.3 | NP_083640.1 | 10 - 11 | 1281 | 74 | |
| GenBank mRNA | AK030773.1 | - | 10 - 11 | 1280 | 74 | |
| AK049162.1 | - | 10 - 11 | 1282 | 74 | ||
| AK054046.1 | - | 10 - 11 | 1281 | 74 | ||
| AK083597.1 | - | 10 - 11 | 1259 | 74 | ||
| AK149863.1 | - | 10 - 11 | 1266 | 74 | ||
| AK151037.1 | - | 8 - 9 | 903 | 74 | ||
| AK159562.1 | - | 10 - 11 | 1248 | 74 | ||
| AK165180.1 | - | 10 - 11 | 1261 | 74 | ||
| AK165917.1 | - | 10 - 11 | 1231 | 74 | ||
| AK169485.1 | - | 10 - 11 | 1263 | 74 | ||
| AK170791.1 | - | 10 - 11 | 1277 | 74 | ||
| AK192137.1 | - | 2 - 3 | 134 | 74 | ||
| AK195315.1 | - | 2 - 3 | 133 | 74 | ||
| AK195952.1 | - | 2 - 3 | 133 | 74 | ||
| AK197098.1 | - | 3 - 4 | 249 | 74 | ||
| AK198268.1 | - | 2 - 3 | 134 | 74 | ||
| AK198818.1 | - | 3 - 4 | 249 | 74 | ||
| AK201813.1 | - | 3 - 4 | 252 | 74 | ||
| AK205650.1 | - | 2 - 3 | 134 | 74 | ||
| BC055328.1 | - | 10 - 11 | 1275 | 6400020 | 74 | |
| BC059047.1 | - | 2 - 3 | 435 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gns
Genomic Map