Assay Details
| Gene Symbol: Gns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029364.3 | NP_083640.1 | 1 - 2 | 272 | 92 | |
| GenBank mRNA | AK030773.1 | - | 1 - 2 | 271 | 92 | |
| AK049162.1 | - | 1 - 2 | 273 | 92 | ||
| AK083597.1 | - | 1 - 2 | 250 | 92 | ||
| AK087265.1 | - | 1 - 2 | 256 | 92 | ||
| AK159562.1 | - | 1 - 2 | 239 | 92 | ||
| AK165180.1 | - | 1 - 2 | 252 | 92 | ||
| AK165917.1 | - | 1 - 2 | 254 | 92 | ||
| AK169485.1 | - | 1 - 2 | 254 | 92 | ||
| AK170791.1 | - | 1 - 2 | 268 | 92 | ||
| BC055328.1 | - | 1 - 2 | 266 | 6400020 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gns
Genomic Map