Assay Details
| Gene Symbol: Gns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029364.3 | NP_083640.1 | 2 - 3 | 332 | 98 | |
| GenBank mRNA | AK030773.1 | - | 2 - 3 | 331 | 98 | |
| AK049162.1 | - | 2 - 3 | 333 | 98 | ||
| AK083597.1 | - | 2 - 3 | 310 | 98 | ||
| AK159562.1 | - | 2 - 3 | 299 | 98 | ||
| AK165180.1 | - | 2 - 3 | 312 | 98 | ||
| AK169485.1 | - | 2 - 3 | 314 | 98 | ||
| AK170791.1 | - | 2 - 3 | 328 | 98 | ||
| AK197322.1 | - | 2 - 3 | 112 | 98 | ||
| BC055328.1 | - | 2 - 3 | 326 | 6400020 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gns
Genomic Map