Assay Details
| Gene Symbol: Gns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029364.3 | NP_083640.1 | 5 - 6 | 701 | 75 | |
| GenBank mRNA | AK030773.1 | - | 5 - 6 | 700 | 75 | |
| AK049162.1 | - | 5 - 6 | 702 | 75 | ||
| AK054046.1 | - | 5 - 6 | 701 | 75 | ||
| AK083597.1 | - | 5 - 6 | 679 | 75 | ||
| AK149863.1 | - | 5 - 6 | 686 | 75 | ||
| AK151037.1 | - | 3 - 4 | 323 | 75 | ||
| AK159562.1 | - | 5 - 6 | 668 | 75 | ||
| AK165180.1 | - | 5 - 6 | 681 | 75 | ||
| AK165917.1 | - | 5 - 6 | 651 | 75 | ||
| AK169485.1 | - | 5 - 6 | 683 | 75 | ||
| AK170791.1 | - | 5 - 6 | 697 | 75 | ||
| BC055328.1 | - | 5 - 6 | 695 | 6400020 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gns
Genomic Map