Assay Details
| Gene Symbol: Gns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029364.3 | NP_083640.1 | 8 - 9 | 1073 | 87 | |
| GenBank mRNA | AK030773.1 | - | 8 - 9 | 1072 | 87 | |
| AK049162.1 | - | 8 - 9 | 1074 | 87 | ||
| AK054046.1 | - | 8 - 9 | 1073 | 87 | ||
| AK149863.1 | - | 8 - 9 | 1058 | 87 | ||
| AK151037.1 | - | 6 - 7 | 695 | 87 | ||
| AK159562.1 | - | 8 - 9 | 1040 | 87 | ||
| AK165180.1 | - | 8 - 9 | 1053 | 87 | ||
| AK165917.1 | - | 8 - 9 | 1023 | 87 | ||
| AK169485.1 | - | 8 - 9 | 1055 | 87 | ||
| AK170791.1 | - | 8 - 9 | 1069 | 87 | ||
| BC055328.1 | - | 8 - 9 | 1067 | 6400020 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gns
Genomic Map