Assay Details
| Gene Symbol: Gns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029364.3 | NP_083640.1 | 9 - 10 | 1178 | 61 | |
| GenBank mRNA | AK030773.1 | - | 9 - 10 | 1177 | 61 | |
| AK049162.1 | - | 9 - 10 | 1179 | 61 | ||
| AK054046.1 | - | 9 - 10 | 1178 | 61 | ||
| AK083597.1 | - | 9 - 10 | 1156 | 61 | ||
| AK149863.1 | - | 9 - 10 | 1163 | 61 | ||
| AK151037.1 | - | 7 - 8 | 800 | 61 | ||
| AK159562.1 | - | 9 - 10 | 1145 | 61 | ||
| AK165180.1 | - | 9 - 10 | 1158 | 61 | ||
| AK165917.1 | - | 9 - 10 | 1128 | 61 | ||
| AK169485.1 | - | 9 - 10 | 1160 | 61 | ||
| AK170791.1 | - | 9 - 10 | 1174 | 61 | ||
| AK197098.1 | - | 2 - 3 | 146 | 61 | ||
| AK198818.1 | - | 2 - 3 | 146 | 61 | ||
| AK201813.1 | - | 2 - 3 | 148 | 61 | ||
| AK208382.1 | - | 2 - 3 | 146 | 61 | ||
| BC055328.1 | - | 9 - 10 | 1172 | 6400020 | 61 | |
| BC059047.1 | - | 1 - 2 | 332 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC059047.1 transcripts.
Set Membership: |
|

Gns
Genomic Map