Assay Details
| Gene Symbol: Tbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019786.4 | NP_062760.3 | 11 - 12 | 1499 | 86 | |
| GenBank mRNA | AF145705.1 | - | 10 - 11 | 1351 | 86 | |
| AF191839.1 | - | 10 - 11 | 1378 | 86 | ||
| AK004649.2 | - | 11 - 12 | 1453 | 86 | ||
| AK083214.1 | - | 11 - 12 | 1447 | 86 | ||
| AK168193.1 | - | 11 - 12 | 1481 | 86 | ||
| BC129910.1 | - | 10 - 11 | 1411 | 40111752 | 86 | |
| BC129911.1 | - | 10 - 11 | 1411 | 40111754 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbk1
Genomic Map