Assay Details
| Gene Symbol: Tbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019786.4 | NP_062760.3 | 18 - 19 | 2120 | 77 | |
| GenBank mRNA | AF145705.1 | - | 17 - 18 | 1972 | 77 | |
| AF191839.1 | - | 17 - 18 | 1999 | 77 | ||
| AK004269.1 | - | 2 - 3 | 182 | 77 | ||
| AK004649.2 | - | 18 - 19 | 2074 | 77 | ||
| AK083214.1 | - | 18 - 19 | 2068 | 77 | ||
| AK168193.1 | - | 18 - 19 | 2102 | 77 | ||
| BC129910.1 | - | 17 - 18 | 2032 | 40111752 | 77 | |
| BC129911.1 | - | 17 - 18 | 2032 | 40111754 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbk1
Genomic Map