Assay Details
| Gene Symbol: Tbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019786.4 | NP_062760.3 | 3 - 4 | 382 | 138 | |
| GenBank mRNA | AF145705.1 | - | 2 - 3 | 234 | 138 | |
| AF191839.1 | - | 2 - 3 | 261 | 138 | ||
| AK004649.2 | - | 3 - 4 | 336 | 138 | ||
| AK083214.1 | - | 3 - 4 | 329 | 138 | ||
| AK168193.1 | - | 3 - 4 | 368 | 138 | ||
| BC129910.1 | - | 2 - 3 | 294 | 40111752 | 138 | |
| BC129911.1 | - | 2 - 3 | 294 | 40111754 | 138 | |
| BU708441.1 | - | 3 - 4 | 375 | 138 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK168193.1 transcripts.
Set Membership: |
|

Tbk1
Genomic Map