Assay Details
| Gene Symbol: Tbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019786.4 | NP_062760.3 | 6 - 7 | 857 | 112 | |
| GenBank mRNA | AF145705.1 | - | 5 - 6 | 709 | 112 | |
| AF191839.1 | - | 5 - 6 | 736 | 112 | ||
| AK004649.2 | - | 6 - 7 | 811 | 112 | ||
| AK083214.1 | - | 6 - 7 | 805 | 113 | ||
| AK168193.1 | - | 6 - 7 | 839 | 112 | ||
| BC129910.1 | - | 5 - 6 | 769 | 40111752 | 112 | |
| BC129911.1 | - | 5 - 6 | 769 | 40111754 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbk1
Genomic Map