Assay Details
| Gene Symbol: Wee2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201370.2 | NP_958758.2 | 2 - 3 | 691 | 106 | |
| GenBank mRNA | AK139573.1 | - | 2 - 3 | 650 | 106 | |
| AK143336.1 | - | 2 - 3 | 615 | 106 | ||
| AK163344.1 | - | 2 - 3 | 693 | 106 | ||
| BC052883.1 | - | 2 - 3 | 598 | 30053490 | 106 | |
| BC080784.1 | - | 2 - 3 | 616 | 30481521 | 106 | |
| DQ011691.1 | - | 2 - 3 | 512 | 106 | ||
More Information
Set Membership: |
|

Wee2
Genomic Map