Assay Details
| Gene Symbol: Wee2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201370.2 | NP_958758.2 | 7 - 8 | 1378 | 71 | |
| GenBank mRNA | AK139573.1 | - | 7 - 8 | 1337 | 71 | |
| AK143336.1 | - | 7 - 8 | 1302 | 71 | ||
| AK163344.1 | - | 7 - 8 | 1380 | 71 | ||
| BC052883.1 | - | 7 - 8 | 1285 | 30053490 | 71 | |
| BC080784.1 | - | 7 - 8 | 1303 | 30481521 | 71 | |
| DQ011691.1 | - | 7 - 8 | 1199 | 71 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Wee2
Genomic Map