Assay Details
| Gene Symbol: Wee2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201370.2 | NP_958758.2 | 9 - 10 | 1570 | 131 | |
| GenBank mRNA | AK139573.1 | - | 9 - 10 | 1529 | 131 | |
| AK143336.1 | - | 9 - 10 | 1494 | 131 | ||
| AK163344.1 | - | 9 - 10 | 1572 | 131 | ||
| BC052883.1 | - | 9 - 10 | 1477 | 30053490 | 131 | |
| BC080784.1 | - | 9 - 10 | 1495 | 30481521 | 131 | |
| DQ011691.1 | - | 9 - 10 | 1391 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wee2
Genomic Map