Assay Details
| Gene Symbol: Map3k15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163085.2 | NP_001156557.2 | 18 - 19 | 2443 | 84 | |
| GenBank mRNA | BC031147.1 | - | 17 - 18 | 1948 | 84 | |
| BC043943.1 | - | 17 - 18 | 1948 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map3k15
Genomic Map