Assay Details
| Gene Symbol: Map3k15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163085.2 | NP_001156557.2 | 7 - 8 | 1183 | 96 | |
| GenBank mRNA | BC031147.1 | - | 6 - 7 | 689 | 96 | |
| BC043943.1 | - | 6 - 7 | 689 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map3k15
Genomic Map