Assay Details
| Gene Symbol: Met | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008591.2 | NP_032617.2 | 17 - 18 | 3685 | 87 | |
| GenBank mRNA | BU514228.1 | - | 3 - 4 | 342 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Met
Genomic Map