Assay Details
| Gene Symbol: Met | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008591.2 | NP_032617.2 | 18 - 19 | 3873 | 85 | |
| GenBank mRNA | BC117969.1 | - | 16 - 17 | 3517 | 85 | |
| BC117970.1 | - | 16 - 17 | 3513 | 85 | ||
| BU514228.1 | - | 4 - 5 | 530 | 85 | ||
| Y00671.1 | - | 16 - 17 | 3518 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Met
Genomic Map