Assay Details
| Gene Symbol: Capza2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007604.2 | NP_031630.1 | 8 - 9 | 679 | 154 | |
| GenBank mRNA | AK007550.1 | - | 8 - 9 | 678 | 154 | |
| AK008204.1 | - | 8 - 9 | 665 | 154 | ||
| AK150751.1 | - | 8 - 9 | 746 | 154 | ||
| AK152674.1 | - | 5 - 6 | 473 | 154 | ||
| AY544984.1 | - | 8 - 9 | 675 | 154 | ||
| BC082589.1 | - | 8 - 9 | 679 | 30626416 | 154 | |
| U16741.1 | - | 8 - 9 | 760 | 154 | ||
More Information
Set Membership: |
|

Capza2
Genomic Map