Assay Details
| Gene Symbol: Hepacam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178899.5 | NP_849230.1 | 1 - 2 | 100 | 71 | |
| GenBank mRNA | AK033728.1 | - | 1 - 2 | 102 | 71 | |
| AK050501.1 | - | 1 - 2 | 101 | 71 | ||
| BC096374.1 | - | 1 - 2 | 87 | 4483627 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hepacam2
Genomic Map