Assay Details
| Gene Symbol: Calcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007588.2 | NP_031614.2 | 9 - 10 | 1246 | 81 | |
| GenBank mRNA | AK039161.1 | - | 9 - 10 | 1191 | 81 | |
| AK162391.1 | - | 9 - 10 | 1001 | 81 | ||
| BC119232.1 | - | 7 - 8 | 753 | 8733981 | 81 | |
| BC119272.1 | - | 7 - 8 | 753 | 8734021 | 81 | |
| U18542.1 | - | 9 - 10 | 1148 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Calcr
Genomic Map