Assay Details
| Gene Symbol: Xpot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081056.1 | NP_001074525.1 | 7 - 8 | 959 | 81 | |
| GenBank mRNA | AK029113.1 | - | 2 - 3 | 412 | 81 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Xpot
Genomic Map